2-Fluoro-N-1-,N-1-dimethyl-1,4-benzenediamine

Supplier: Matrix Scientific

051324-500MG 051324-2.5G
101895-188EA 364.62 USD
101895-188 101895-190
2-Fluoro-N-1-,N-1-dimethyl-1,4-benzenediamine
2-Fluoro-N-1-,N-1-dimethyl-1,4-benzenediamine

Hazard: Irritant

Formula: C₈H₁₁FN₂
MW: 154.19 g/mol
MDL Number: MFCD10694806
CAS Number: 3824-31-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR