4-[3,4-Dihydro-1(2H)-quinolinyl]-3-methylaniline
Supplier: Matrix Scientific
051321-500MG
051321-2.5G
101895-176EA
364.62
USD
101895-176
101895-178
4-[3,4-Dihydro-1(2H)-quinolinyl]-3-methylaniline
4-[3,4-Dihydro-1(2H)-quinolinyl]-3-methylaniline
Hazard: Irritant
Formula:
C₁₆H₁₈N₂ |
MDL Number:
MFCD11214002 CAS Number: 1094746-54-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...