N-1-Cyclohexyl-N-1-ethyl-2-methyl-1,4-benzenediamine

Supplier: Matrix Scientific

051317-500MG 051317-2.5G
101895-160EA 364.62 USD
101895-160 101895-162
N-1-Cyclohexyl-N-1-ethyl-2-methyl-1,4-benzenediamine
N-1-Cyclohexyl-N-1-ethyl-2-methyl-1,4-benzenediamine

Hazard: Irritant

Formula: C₁₅H₂₄N₂
MDL Number: MFCD11214006
CAS Number: 1094746-55-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR