N-1-Benzyl-N-1,2-dimethyl-1,4-benzenediamine

Supplier: Matrix Scientific

Synonyms: N1-Benzyl-N1,2-dimethylbenzene-1,4-diamine

051316-500MG 051316-2.5G
101895-156EA 364.62 USD
101895-156 101895-158
N-1-Benzyl-N-1,2-dimethyl-1,4-benzenediamine
N-1-Benzyl-N-1,2-dimethyl-1,4-benzenediamine

Hazard: Irritant

Formula: C₁₅H₁₈N₂
MW: 226.32 g/mol
MDL Number: MFCD11214012
CAS Number: 1094646-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR