N-1-Cyclohexyl-N-1,2-dimethyl-1,4-benzenediamine

Supplier: Matrix Scientific

051314-500MG 051314-2.5G
101895-148EA 364.62 USD
101895-148 101895-150
N-1-Cyclohexyl-N-1,2-dimethyl-1,4-benzenediamine
N-1-Cyclohexyl-N-1,2-dimethyl-1,4-benzenediamine

Hazard: Irritant

Formula: C₁₄H₂₂N₂
MW: 218.34 g/mol
MDL Number: MFCD11214003
CAS Number: 1094497-09-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR