1-(4-Amino-2-methylphenyl)-4-piperidinol
Supplier: Matrix Scientific
051312-500MG
051312-2.5G
101895-140EA
364.62
USD
101895-140
101895-142
1-(4-Amino-2-methylphenyl)-4-piperidinol
1-(4-Amino-2-methylphenyl)-4-piperidinol
Hazard: Irritant
Formula:
C₁₂H₁₈N₂O |
MDL Number:
MFCD11214000 CAS Number: 873537-60-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...