4-(1-Azepanyl)-3-methylaniline

Supplier: Matrix Scientific

051310-500MG 051310-2.5G
101895-132EA 364.62 USD
101895-132 101895-134
4-(1-Azepanyl)-3-methylaniline
4-(1-Azepanyl)-3-methylaniline

Hazard: Irritant

Formula: C₁₃H₂₀N₂
MDL Number: MFCD11213995
CAS Number: 1094646-23-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR