4-(4-Benzyl-1-piperazinyl)-3-methylphenylamine

Supplier: Matrix Scientific

051309-500MG 051309-2.5G
101895-128EA 364.62 USD
101895-128 101895-130
4-(4-Benzyl-1-piperazinyl)-3-methylphenylamine
4-(4-Benzyl-1-piperazinyl)-3-methylphenylamine

Hazard: Irritant

Formula: C₁₈H₂₃N₃
MDL Number: MFCD13561474
CAS Number: 1097161-95-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR