3-Methyl-4-(4-methyl-1-piperazinyl)aniline

Supplier: Matrix Scientific

051308-500MG 051308-2.5G
101895-124EA 364.62 USD
101895-124 101895-126
3-Methyl-4-(4-methyl-1-piperazinyl)aniline
3-Methyl-4-(4-methyl-1-piperazinyl)aniline

Hazard: Irritant

Formula: C₁₂H₁₉N₃
MDL Number: MFCD08444067
CAS Number: 681004-50-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR