3-Methyl-4-(4-methyl-1-piperidinyl)aniline

Supplier: Matrix Scientific

051307-500MG 051307-2.5G
101895-120EA 461.54 USD
101895-120 101895-122
3-Methyl-4-(4-methyl-1-piperidinyl)aniline
3-Methyl-4-(4-methyl-1-piperidinyl)aniline

Hazard: Irritant

Formula: C₁₃H₂₀N₂
MDL Number: MFCD11106381
CAS Number: 1152822-12-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR