N-1,N-1-Diallyl-2-methyl-1,4-benzenediamine
Supplier: Matrix Scientific
051304-500MG
051304-2.5G
101895-108EA
364.62
USD
101895-108
101895-110
N-1,N-1-Diallyl-2-methyl-1,4-benzenediamine
N-1,N-1-Diallyl-2-methyl-1,4-benzenediamine
Hazard: Irritant
Formula:
C₁₃H₁₈N₂ |
MDL Number:
MFCD13561473 CAS Number: 138578-90-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...