2-[4-Amino(ethyl)-2-methylanilino]-1-ethanol

Supplier: Matrix Scientific

051303-500MG 051303-2.5G
101895-104EA 364.62 USD
101895-104 101895-106
2-[4-Amino(ethyl)-2-methylanilino]-1-ethanol
2-[4-Amino(ethyl)-2-methylanilino]-1-ethanol

Hazard: Irritant

Formula: C₁₁H₁₈N₂O
MDL Number: MFCD12813173
CAS Number: 72225-29-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR