N-1-Butyl-N-1,(2-dimethyl)-1,4-benzenediamine

Supplier: Matrix Scientific

051302-500MG 051302-2.5G
101895-100EA 364.62 USD
101895-100 101895-102
N-1-Butyl-N-1,(2-dimethyl)-1,4-benzenediamine
N-1-Butyl-N-1,(2-dimethyl)-1,4-benzenediamine

Hazard: Irritant

Formula: C₁₂H₂₀N₂
MDL Number: MFCD11214013
CAS Number: 1094544-25-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR