1-(4-Amino-2-methylphenyl)-3-pyrrolidinol

Supplier: Matrix Scientific

051301-500MG 051301-2.5G
101895-096EA 364.62 USD
101895-096 101895-098
1-(4-Amino-2-methylphenyl)-3-pyrrolidinol
1-(4-Amino-2-methylphenyl)-3-pyrrolidinol

Hazard: Irritant

Formula: C₁₁H₁₆N₂O
MDL Number: MFCD13561472
CAS Number: 503457-33-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR