3-Methyl-4-(4-morpholinyl)aniline

Supplier: Matrix Scientific

Synonyms: 3-Methyl-4-morpholinoaniline

051300-500MG 051300-2.5G
101895-092EA 189.23 USD
101895-092 101895-094
3-Methyl-4-(4-morpholinyl)aniline
3-Methyl-4-(4-morpholinyl)aniline

Hazard: Irritant

Formula: C₁₁H₁₆N₂O
MW: 192.26 g/mol
MDL Number: MFCD09755865
CAS Number: 112900-82-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR