3-Methyl-4-(1-pyrrolidinyl)aniline

Supplier: Matrix Scientific

051296-500MG 051296-2.5G
101895-076EA 364.62 USD
101895-076 101895-078
3-Methyl-4-(1-pyrrolidinyl)aniline
3-Methyl-4-(1-pyrrolidinyl)aniline

Hazard: Irritant

Formula: C₁₁H₁₆N₂
MDL Number: MFCD06336762
CAS Number: 16089-43-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR