4-(3,4-Dihydroisoquinolin-2(1H)-yl)-3-(trifluoromethyl)aniline

Supplier: Matrix Scientific

051290-500MG 051290-2.5G
101895-052EA 364.62 USD
101895-052 101895-054
4-(3,4-Dihydroisoquinolin-2(1H)-yl)-3-(trifluoromethyl)aniline
4-(3,4-Dihydroisoquinolin-2(1H)-yl)-3-(trifluoromethyl)aniline

Hazard: Irritant

Formula: C₁₆H₁₅F₃N₂
CAS Number: 914348-87-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR