2-{4-[4-Amino-2-(trifluoromethyl)phenyl]-1-piperazinyl}-1-ethanol

Supplier: Matrix Scientific

051289-500MG 051289-2.5G
101895-048EA 461.54 USD
101895-048 101895-050
2-{4-[4-Amino-2-(trifluoromethyl)phenyl]-1-piperazinyl}-1-ethanol
2-{4-[4-Amino-2-(trifluoromethyl)phenyl]-1-piperazinyl}-1-ethanol

Hazard: Irritant

Formula: C₁₃H₁₈F₃N₃O
MDL Number: MFCD13182433
CAS Number: 1183251-84-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR