N-1-Cyclohexyl-N-1-ethyl-2-(trifluoromethyl)-1,4-benzenediamine

Supplier: Matrix Scientific

051288-500MG 051288-2.5G
101895-044EA 364.62 USD
101895-044 101895-046
N-1-Cyclohexyl-N-1-ethyl-2-(trifluoromethyl)-1,4-benzenediamine
N-1-Cyclohexyl-N-1-ethyl-2-(trifluoromethyl)-1,4-benzenediamine

Hazard: Irritant

Formula: C₁₅H₂₁F₃N₂
MDL Number: MFCD13561467
CAS Number: 1220016-64-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR