4-(2-Ethyl-1-piperidinyl)-3-(trifluoromethyl)phenylamine

Supplier: Matrix Scientific

051286-500MG 051286-2.5G
101895-036EA 364.62 USD
101895-036 101895-038
4-(2-Ethyl-1-piperidinyl)-3-(trifluoromethyl)phenylamine
4-(2-Ethyl-1-piperidinyl)-3-(trifluoromethyl)phenylamine

Hazard: Irritant

Formula: C₁₄H₁₉F₃N₂
MDL Number: MFCD13235307
CAS Number: 946669-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR