N-1-Cyclohexyl-N-1-methyl-2-(trifluoromethyl)-1,4-benzenediamine
Supplier: Matrix Scientific
051285-500MG
051285-2.5G
101895-032EA
364.62
USD
101895-032
101895-034
N-1-Cyclohexyl-N-1-methyl-2-(trifluoromethyl)-1,4-benzenediamine
N-1-Cyclohexyl-N-1-methyl-2-(trifluoromethyl)-1,4-benzenediamine
Hazard: Irritant
Formula:
C₁₄H₁₉F₃N₂ |
MDL Number:
MFCD12876417 CAS Number: 1183349-95-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...