N-1,N-1-Dipropyl-2-(trifluoromethyl)-1,4-benzenediamine
Supplier: Matrix Scientific
051284-500MG
051284-2.5G
101895-028EA
364.62
USD
101895-028
101895-030
N-1,N-1-Dipropyl-2-(trifluoromethyl)-1,4-benzenediamine
N-1,N-1-Dipropyl-2-(trifluoromethyl)-1,4-benzenediamine
Hazard: Irritant
Formula:
C₁₃H₁₉F₃N₂ MW: 260.3 g/mol |
MDL Number:
MFCD12876416 CAS Number: 25903-26-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...