4-[4-Amino-2-(trifluoromethyl)phenyl]-2-piperazinone

Supplier: Matrix Scientific

051282-500MG 051282-2.5G
101895-020EA 364.62 USD
101895-020 101895-022
4-[4-Amino-2-(trifluoromethyl)phenyl]-2-piperazinone
4-[4-Amino-2-(trifluoromethyl)phenyl]-2-piperazinone

Hazard: Irritant

Formula: C₁₁H₁₂F₃N₃O
MDL Number: MFCD12876415
CAS Number: 1183065-10-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR