4-(3-Methyl-1-piperidinyl)-3-(trifluoromethyl)phenylamine
Supplier: Matrix Scientific
051278-500MG
051278-2.5G
101895-004EA
364.62
USD
101895-004
101895-006
4-(3-Methyl-1-piperidinyl)-3-(trifluoromethyl)phenylamine
4-(3-Methyl-1-piperidinyl)-3-(trifluoromethyl)phenylamine
Hazard: Irritant
Formula:
C₁₃H₁₇F₃N₂ |
MDL Number:
MFCD13235306 CAS Number: 946730-96-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...