4-(2-Methyl-1-piperidinyl)-3-(trifluoromethyl)phenylamine

Supplier: Matrix Scientific

051277-500MG 051277-2.5G
101895-000EA 364.62 USD
101895-000 101895-002
4-(2-Methyl-1-piperidinyl)-3-(trifluoromethyl)phenylamine
4-(2-Methyl-1-piperidinyl)-3-(trifluoromethyl)phenylamine

Hazard: Irritant

Formula: C₁₃H₁₇F₃N₂
MDL Number: MFCD13204851
CAS Number: 946719-19-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR