N-1,N-1-Diallyl-2-(trifluoromethyl)-1,4-benzenediamine
Supplier: Matrix Scientific
051276-500MG
051276-2.5G
101894-996EA
364.62
USD
101894-996
101894-998
N-1,N-1-Diallyl-2-(trifluoromethyl)-1,4-benzenediamine
N-1,N-1-Diallyl-2-(trifluoromethyl)-1,4-benzenediamine
Hazard: Irritant
Formula:
C₁₃H₁₅F₃N₂ |
MDL Number:
MFCD13561465 CAS Number: 1220016-44-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...