2-[4-Amino(ethyl)-2-(trifluoromethyl)anilino]-1-ethanol
Supplier: Matrix Scientific
051275-500MG
051275-2.5G
101894-992EA
364.62
USD
101894-992
101894-994
2-[4-Amino(ethyl)-2-(trifluoromethyl)anilino]-1-ethanol
2-[4-Amino(ethyl)-2-(trifluoromethyl)anilino]-1-ethanol
Hazard: Irritant
Formula:
C₁₁H₁₅F₃N₂O |
MDL Number:
MFCD12890318 CAS Number: 1184010-61-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...