1-[4-Amino-2-(trifluoromethyl)phenyl]-3-pyrrolidinol
Supplier: Matrix Scientific
051273-500MG
051273-2.5G
101894-984EA
364.62
USD
101894-984
101894-986
1-[4-Amino-2-(trifluoromethyl)phenyl]-3-pyrrolidinol
1-[4-Amino-2-(trifluoromethyl)phenyl]-3-pyrrolidinol
Hazard: Irritant
Formula:
C₁₁H₁₃F₃N₂O |
MDL Number:
MFCD13561464 CAS Number: 1220019-17-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...