N-1,N-1-Diethyl-2-(trifluoromethyl)-1,4-benzenediamine

Supplier: Matrix Scientific

051271-500MG 051271-2.5G
101894-976EA 364.62 USD
101894-976 101894-978
N-1,N-1-Diethyl-2-(trifluoromethyl)-1,4-benzenediamine
N-1,N-1-Diethyl-2-(trifluoromethyl)-1,4-benzenediamine

Hazard: Irritant

Formula: C₁₁H₁₅F₃N₂
MW: 232.25 g/mol
MDL Number: MFCD12762366
CAS Number: 105296-03-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR