2-[4-(4-Aminophenyl)-1-piperazinyl]-1-ethanol

Supplier: Matrix Scientific

051264-500MG 051264-2.5G
101894-948EA 384.62 USD
101894-948 101894-950
2-[4-(4-Aminophenyl)-1-piperazinyl]-1-ethanol
2-[4-(4-Aminophenyl)-1-piperazinyl]-1-ethanol

Hazard: Irritant

Formula: C₁₂H₁₉N₃O
MW: 221.3 g/mol
MDL Number: MFCD00702247
CAS Number: 5521-39-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR