4-(2-Ethyl-1-piperidinyl)aniline

Supplier: Matrix Scientific

051261-500MG 051261-2.5G
101894-936EA 461.54 USD
101894-936 101894-938
4-(2-Ethyl-1-piperidinyl)aniline
4-(2-Ethyl-1-piperidinyl)aniline

Hazard: Irritant

Formula: C₁₃H₂₀N₂
MDL Number: MFCD11618639
CAS Number: 946718-57-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR