N-1-Cyclohexyl-N-1-methyl-1,4-benzenediamine

Supplier: Matrix Scientific

051260-500MG 051260-2.5G
101894-932EA 364.62 USD
101894-932 101894-934
N-1-Cyclohexyl-N-1-methyl-1,4-benzenediamine
N-1-Cyclohexyl-N-1-methyl-1,4-benzenediamine

Hazard: Irritant

Formula: C₁₃H₂₀N₂
MW: 204.32 g/mol
MDL Number: MFCD07365103
CAS Number: 93087-80-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR