4-(1-Azepanyl)aniline

Supplier: Matrix Scientific

051256-500MG 051256-2.5G
101894-916EA 364.62 USD
101894-916 101894-918
4-(1-Azepanyl)aniline
4-(1-Azepanyl)aniline

Hazard: Irritant

Formula: C₁₂H₁₈N₂
MW: 190.29 g/mol
MDL Number: MFCD01536453
CAS Number: 57356-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR