N-1,N-1-Diallyl-1,4-benzenediamine

Supplier: Matrix Scientific

051255-500MG 051255-2.5G
101894-912EA 364.62 USD
101894-912 101894-914
N-1,N-1-Diallyl-1,4-benzenediamine
N-1,N-1-Diallyl-1,4-benzenediamine

Hazard: Irritant

Formula: C₁₂H₁₆N₂
MW: 188.27 g/mol
MDL Number: MFCD13561462
CAS Number: 74332-66-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR