2-[4-Amino(ethyl)anilino]-1-ethanol

Supplier: Matrix Scientific

051254-500MG 051254-2.5G
101894-908EA 364.62 USD
101894-908 101894-910
2-[4-Amino(ethyl)anilino]-1-ethanol
2-[4-Amino(ethyl)anilino]-1-ethanol

Hazard: Irritant

Formula: C₁₀H₁₆N₂O
MDL Number: MFCD01006065
CAS Number: 92-65-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR