2-[4-Amino(methyl)anilino]-1-ethanol
Supplier: Matrix Scientific
Synonyms:
2-[(4-Aminophenyl)(methyl)amino]ethan-1-ol
051250-500MG
051250-2.5G
101894-892EA
364.62
USD
101894-892
101894-894
2-[4-Amino(methyl)anilino]-1-ethanol
2-[4-Amino(methyl)anilino]-1-ethanol
Hazard: Irritant
Formula:
C₉H₁₄N₂O MW: 166.22 g/mol |
MDL Number:
MFCD11184967 CAS Number: 17625-86-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...