3-Chloro-4-[3,4-dihydro-1(2H)-quinolinyl]aniline

Supplier: Matrix Scientific

051245-500MG 051245-2.5G
101894-872EA 364.62 USD
101894-872 101894-874
3-Chloro-4-[3,4-dihydro-1(2H)-quinolinyl]aniline
3-Chloro-4-[3,4-dihydro-1(2H)-quinolinyl]aniline

Hazard: Irritant

Formula: C₁₅H₁₅ClN₂
MDL Number: MFCD08699354
CAS Number: 937608-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR