3-Chloro-4-(2,3-dihydro-1H-indol-1-yl)aniline

Supplier: Matrix Scientific

051244-500MG 051244-2.5G
101894-868EA 364.62 USD
101894-868 101894-870
3-Chloro-4-(2,3-dihydro-1H-indol-1-yl)aniline
3-Chloro-4-(2,3-dihydro-1H-indol-1-yl)aniline

Hazard: Irritant

Formula: C₁₄H₁₃ClN₂
MDL Number: MFCD08699353
CAS Number: 937608-38-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR