3-Chloro-4-[3,4-dihydro-2(1H)-isoquinolinyl]aniline

Supplier: Matrix Scientific

051243-500MG 051243-2.5G
101894-864EA 364.62 USD
101894-864 101894-866
3-Chloro-4-[3,4-dihydro-2(1H)-isoquinolinyl]aniline
3-Chloro-4-[3,4-dihydro-2(1H)-isoquinolinyl]aniline

Hazard: Irritant

Formula: C₁₅H₁₅ClN₂
MW: 258.75 g/mol
MDL Number: MFCD07365138
CAS Number: 893750-76-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR