N-(4-Amino-2-chlorophenyl)-N-cyclohexyl-N-ethylamine

Supplier: Matrix Scientific

051242-500MG 051242-2.5G
101894-860EA 364.62 USD
101894-860 101894-862
N-(4-Amino-2-chlorophenyl)-N-cyclohexyl-N-ethylamine
N-(4-Amino-2-chlorophenyl)-N-cyclohexyl-N-ethylamine

Hazard: Irritant

Formula: C₁₄H₂₁ClN₂
MDL Number: MFCD11118544
CAS Number: 1036446-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR