3-Chloro-4-(2-ethyl-1-piperidinyl)aniline

Supplier: Matrix Scientific

051240-500MG 051240-2.5G
101894-852EA 364.62 USD
101894-852 101894-854
3-Chloro-4-(2-ethyl-1-piperidinyl)aniline
3-Chloro-4-(2-ethyl-1-piperidinyl)aniline

Hazard: Irritant

Formula: C₁₃H₁₉ClN₂
MDL Number: MFCD11618642
CAS Number: 946668-90-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR