N-(4-Amino-2-chlorophenyl)-N-cyclohexyl-N-methylamine
Supplier: Matrix Scientific
051239-500MG
051239-2.5G
101894-848EA
364.62
USD
101894-848
101894-850
N-(4-Amino-2-chlorophenyl)-N-cyclohexyl-N-methylamine
N-(4-Amino-2-chlorophenyl)-N-cyclohexyl-N-methylamine
Hazard: Irritant
Formula:
C₁₃H₁₉ClN₂ |
MDL Number:
MFCD07365141 CAS Number: 85896-15-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...