1-(4-Amino-2-chlorophenyl)-4-piperidinol

Supplier: Matrix Scientific

051237-500MG 051237-2.5G
101894-840EA 364.62 USD
101894-840 101894-842
1-(4-Amino-2-chlorophenyl)-4-piperidinol
1-(4-Amino-2-chlorophenyl)-4-piperidinol

Hazard: Irritant

Formula: C₁₁H₁₅ClN₂O
MDL Number: MFCD09734047
CAS Number: 953743-03-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR