4-(4-Amino-2-chlorophenyl)-2-piperazinone

Supplier: Matrix Scientific

051236-500MG 051236-2.5G
101894-836EA 364.62 USD
101894-836 101894-838
4-(4-Amino-2-chlorophenyl)-2-piperazinone
4-(4-Amino-2-chlorophenyl)-2-piperazinone

Hazard: Irritant

Formula: C₁₀H₁₂ClN₃O
MW: 225.68 g/mol
MDL Number: MFCD09043886
CAS Number: 926250-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR