4-(1-Azepanyl)-3-chloroaniline

Supplier: Matrix Scientific

051235-500MG 051235-2.5G
101894-832EA 364.62 USD
101894-832 101894-834
4-(1-Azepanyl)-3-chloroaniline
4-(1-Azepanyl)-3-chloroaniline

Hazard: Irritant

Formula: C₁₂H₁₇ClN₂
MW: 224.74 g/mol
MDL Number: MFCD08691842
CAS Number: 915921-17-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR