4-(4-Benzyl-1-piperazinyl)-3-chlorophenylamine

Supplier: Matrix Scientific

051234-500MG 051234-2.5G
101894-828EA 364.62 USD
101894-828 101894-830
4-(4-Benzyl-1-piperazinyl)-3-chlorophenylamine
4-(4-Benzyl-1-piperazinyl)-3-chlorophenylamine

Hazard: Irritant

Formula: C₁₇H₂₀ClN₃
MDL Number: MFCD07365136
CAS Number: 55403-32-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR