3-Chloro-4-(3-methyl-1-piperidinyl)aniline

Supplier: Matrix Scientific

051233-500MG 051233-2.5G
101894-824EA 364.62 USD
101894-824 101894-826
3-Chloro-4-(3-methyl-1-piperidinyl)aniline
3-Chloro-4-(3-methyl-1-piperidinyl)aniline

Hazard: Irritant

Formula: C₁₂H₁₇ClN₂
MDL Number: MFCD07365135
CAS Number: 893750-70-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR