N-1-,N-1-Diallyl-2-chloro-1,4-benzenediamine
Supplier: Matrix Scientific
051231-500MG
051231-2.5G
101894-816EA
364.62
USD
101894-816
101894-818
N-1-,N-1-Diallyl-2-chloro-1,4-benzenediamine
N-1-,N-1-Diallyl-2-chloro-1,4-benzenediamine
Hazard: Irritant
Formula:
C₁₂H₁₅ClN₂ |
MDL Number:
MFCD13561460 CAS Number: 1219957-06-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...