3-Chloro-4-(1-pyrrolidinyl)aniline

Supplier: Matrix Scientific

051225-500MG 051225-2.5G
101894-792EA 461.54 USD
101894-792 101894-794
3-Chloro-4-(1-pyrrolidinyl)aniline
3-Chloro-4-(1-pyrrolidinyl)aniline

Hazard: Irritant

Formula: C₁₀H₁₃ClN₂
MDL Number: MFCD02628414
CAS Number: 16089-44-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR