2-[3,4-Dihydro-1(2H)-quinolinyl]-5-fluoroaniline

Supplier: Matrix Scientific

051221-500MG 051221-2.5G
101894-776EA 364.62 USD
101894-776 101894-778
2-[3,4-Dihydro-1(2H)-quinolinyl]-5-fluoroaniline
2-[3,4-Dihydro-1(2H)-quinolinyl]-5-fluoroaniline

Hazard: Irritant

Formula: C₁₅H₁₅FN₂
MW: 242.3 g/mol
MDL Number: MFCD08699503
CAS Number: 937597-69-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR